missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Acetone, specified according to the requirements of Ph.Eur.
Handla allt< titel> Produkter
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
5 L
25 L
Förpackningsstorlek
1 L
25 L
5 L
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Melting Point | -95.0°C |
| Density | 0.7900 g/mL |
| Boiling Point | 56.0°C |
| Quantity | 1 L |
| Flash Point | -20°C |
| Packaging | Glass Bottle |
| Refractive Index | 1.3585 |
| Specific Gravity | 0.79 |
| Linear Formula | CH3COCH3 |
| Viscosity | 0.32 mPa.s (20°C) |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- Acetone
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Acetone
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering