missing translation for 'onlineSavingsMsg'
Learn More
Acetone, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals Product Code.: 11944811

Acetone, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Product Code. 11944811
25 L
Click to view available options
Quantity:
1 L
5 L
25 L
This item is not returnable. View return policy

Product Code. 11944811

Brand: Thermo Scientific Acros 447450250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95.0°C
Density 0.7900 g/mL
Boiling Point 56.0°C
Quantity 25 L
Flash Point -20°C
Packaging Plastic Drum
Refractive Index 1.3585
Specific Gravity 0.79
Linear Formula CH3COCH3
Viscosity 0.32 mPa.s (20°C)
Merck Index 15, 65
Solubility Information Solubility aq. soln.: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Chemical Name or Material Acetone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527023 Certificate TypeCertificate of Analysis Date16/02/2025 Catalog Number447450010,447450250,447450050
Lot Number2524360 Certificate TypeCertificate of Analysis Date16/02/2025 Catalog Number447450010,447450250,447450050
Lot Number2524176 Certificate TypeCertificate of Analysis Date16/02/2025 Catalog Number447450010,447450250,447450050
Lot Number2032701 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number447450010,447450250
Lot Number1903714 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number447450010,447450250
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.