missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Technical, Fisher Chemical™

Product Code. 10652241 Shop All Fisher Chemical Products
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
missing translation for 'unitSize'
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10652241

missing translation for 'mfr': Fisher Chemical A/0520/25

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Packaging Metal can
CAS 67-64-1
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPaS at 20°C
Vapor Density 2
Physical Form Liquid
Molecular Formula C3H6O
MDL Number 8765
Color Colorless
pH 7
Melting Point -95°C
Boiling Point 56°C
Quantity 25 L
Formula Weight 58.08g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Acetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.