missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™

Product Code. 10585635
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8, 76-05-1
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number 4169
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

pH 2
Melting Point -50°C
Absorbance ≤0.03 AU at 254nm, ≤0.4 AU at 230nm, ≤0.55 AU at 220nm, ≤0.6 AU at 210nm
Decomposition Information >500°C
Density 0.7810g/cm³
Boiling Point 82°C
Quantity 2.5 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥97.5 %
Residue after Evaporation ≤1ppm
Linear Formula CH3CN
Water ≤0.01%
Merck Index 15, 9847
Formula Weight 41.04
Percent Purity 0.095 to 0.105% (v/v) Trifluoroacetic Acid
Grade OPTIMA™ LC-MS
Chemical Name or Material 0.1% Trifluoroacetic Acid in Acetonitrile (v/v), Solvent Blends
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile with 0.1% TFA (v/v) for HPLC Gradient analysis
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.