missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ≥99.5% (GC), Honeywell™

≥99.5% (GC)

595.75 € - 2241.96 €

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15625910
View Documents Promotion Details
Honeywell Research Chemicals
60004-6X1L
6 x 1 L Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15695900
View Documents Promotion Details
Honeywell Research Chemicals
60004-4X2.5L
4 x 2.5 L Poly Bottle
1270.16 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15605910
View Documents Promotion Details
Honeywell Research Chemicals
60004-4X5L
4 x 5 L Poly Bottle
2241.96 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-48°C
Colorless
81°C to 82°C
973°C
C2H3N
MFCD00001878
UN1648
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.5% (GC)
75-05-8
0.786g/mL at 25°C
0.16
1.344
CH3CN
72.8mmHg (20°C)
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
Acetonitrile
Videos
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

missing translation for 'dotInformation' : 3 / PGII

missing translation for 'rtecsNumber' : 200-835-2

SDS
Documents

Documents

Product Certifications