missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10193051
Click to view available options
Quantity:
100 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methylkyanid, methane, cyano, methyl cyanide, mecn, methanecarbonitrile, ethanenitrile, ethyl nitrile, cyanure de methyl, cyanomethane, acetonitrile, 100mL, 0.002 % max., Vapor Pressure: 97mbar at 20 deg.C, 99.9+%, 02, 183, 81.0 deg.C to 82.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46.0°C
Decomposition Information >500°C
Density 0.7810g/mL
Boiling Point 81.0°C to 82.0°C
Quantity 100 mL
Flash Point 2°C
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Assay Percent Range 99.9+%
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.781
Linear Formula CH3CN
Vapor Pressure Vapor Pressure: 97mbar at 20°C
Viscosity 0.36 mPa.s (20°C)
Beilstein 02, 183
Water 0.001% Max. (Coulometric)
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Extra Dry
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Acetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'