missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™

Product Code. 15578664 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Flint
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15578664

Brand: Avantor J.T.Baker 985302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cutting-edge LC/MS applications

    For use in LC/MS UHPLC

Specifications

CAS 75-05-8
Quantity 1 L
Flash Point 2.00°C
Packaging Narrow Mouth Flint
Residue after Evaporation ≤0.8ppm
Specific Gravity 1L = 0.79kg
Molecular Formula C2H3N
Water ≤100ppb
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Meets ACS reagent chemical requirements
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1LT ACETONITRILE ULTRA LCMS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis