missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™, ≥99.9% (GC), for LC-NMR, Honeywell Riedel-de Haën™

CHROMASOLV™ NMR, for LC-NMR,≥99.9% (GC)

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15624720
View Documents Promotion Details
Honeywell-Chromasolv
34955-500ML
500 mL N/A
N/A
15614720
View Documents Promotion Details
Honeywell-Chromasolv
34955-1L
1 L N/A
N/A
15644720
View Documents Promotion Details
Honeywell-Chromasolv
34955-6X500ML
6 x 500 mL N/A
N/A
15634720
View Documents Promotion Details
Honeywell-Chromasolv
34955-6X1L
6 x 1 L N/A
N/A
Description

Description

  • Solvents for LC-NMR applications
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

75-05-8
0.786g/mL at 25°C
2°C (35.6°F)
C2H3N
MFCD00001878
UN1648
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9% (GC)
Acetonitrile
Colorless
0.16
973°F
CH3CN
72.8 mmHg (20°C)
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
CHROMASOLV™
Videos
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

missing translation for 'rtecsNumber' : AL7700000

SDS
Documents

Documents

Product Certifications