missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
250 mL
1 L
2.5 L
4 L
20 L
4 x 2.5 L
6 x 1 L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities

Specifications
Specifications
Color | Colorless |
Density | 0.786g/mL at 25°C |
Quantity | 250 mL |
Explosion Limit | 0.16 |
Flash Point | 2°C (35.6°F) |
Ignition Point | 973°F |
Linear Formula | CH3CN |
Vapor Pressure | 72.8 mmHg (20°C) |
UN Number | UN1648 |
Formula Weight | 41.05 |
Show More |
Safety and Handling


Product Identifier
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction