missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15674520
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Quantity 2.5 L
Packaging Glass Bottle
UN Number UN1648
Beilstein 741857
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Acetonitrile, CHROMASOLV, for HPLC, gradient grade, 99.9%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'