missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for DNA Synthesis and Protein Sequencing, Fisher Chemical™
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Amber glass bottle, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
81.25 € - 164.00 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.05 |
MDL Number | 1878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10030010
|
Fisher Chemical
A/0628/15 |
1 L | Amber Glass Bottle | N/A | N/A | N/A | |||||
10510911
|
Fisher Chemical
A/0628/17 |
2.5 L | Amber Glass Bottle | N/A | N/A | N/A | |||||
10151560
|
Fisher Chemical
A/0628/PB17 |
2.5 L | Plastic-coated Glass Bottle |
164.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, Colorless, 41.05g/mol, -46 deg.C, 1878, Amber glass bottle, Liquid, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C
Chemical Identifiers
75-05-8 | |
41.05 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CC#N |
C2H3N | |
1878 | |
acetonitrile |
Specifications
-46°C | |
Colorless | |
Liquid | |
1878 | |
0.36cp at 20°C | |
CC#N | |
41.05 |
75-05-8 | |
81°C | |
C2H3N | |
97mbar at 20°C | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
41.05g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, for DNA Synthesis and Protein Sequencing, Fisher Chemical™