missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile E CHROMASOLV™, for HPLC, for UV, ≥99.9% (GC), Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC, for UV,≥99.9% (GC)
201.97 € - 1682.81 €
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15684680
|
Honeywell-Chromasolv
34888-1L |
1 L |
201.97 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694680
|
Honeywell-Chromasolv
34888-2.5L |
2.5 L |
363.70 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614690
|
Honeywell-Chromasolv
34888-6X1L |
6 x 1 L |
1087.62 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15604690
|
Honeywell-Chromasolv
34888-4X2.5L |
4 x 2.5 L |
1682.81 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
75-05-8 | |
MFCD00001878 | |
6342 | |
acetonitrile |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CHEBI:38472 | |
CC#N |
Specifications
-48°C | |
0.786g/mL at 25°C(lit.) | |
n20/D 1.344 | |
CH3CN | |
72.8mmHg (20°C) | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
≥99.9% (GC) | |
Acetonitrile, for HPLC, for UV |
75-05-8 | |
81°C to 82°C | |
C2H3N | |
MFCD00001878 | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
CHROMASOLV™ |
Safety and Handling
P210-P280-P305 + P351 + P338
H225-H302 + H312 + H332-H319
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title