missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for HPLC, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
Description
C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

Specifications
Specifications
Melting Point | -46°C |
Color | Colorless |
Quantity | 5 L |
Packaging | Aluminum Can |
Vapor Pressure | 97 mbar at 20°C |
Viscosity | 0.36 cp at 20°C |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528119 | Certificate of Analysis | 24/02/2025 | A/0626/27SS,A/0626/99W,A/0626/21,A/0626/PB17X,A/0626/10RSS,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17,A/0626/30RSS |
2528118 | Certificate of Analysis | 24/02/2025 | A/0626/27SS,A/0626/99W,A/0626/21,A/0626/PB17X,A/0626/10RSS,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17,A/0626/30RSS |
2527244 | Certificate of Analysis | 17/02/2025 | A/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17 |
2527243 | Certificate of Analysis | 17/02/2025 | A/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17 |
2526614 | Certificate of Analysis | 12/02/2025 | A/0626/99W,A/0626/10RSS,A/0626/30RSS,A/0626/27SS,A/0626/21,A/0626/PB17,A/0626/PB17X,A/0626/15,A/0626/17X,A/0626/17Z,A/0626/25RSS,A/0626/15X,A/0626/17 |
Safety and Handling


Product Identifier
- Acetonitrile
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, for HPLC, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction