missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, for HPLC, Gradient Grade, Reagent European Pharmacopoeia, Honeywell Burdick & Jackson™

Product Code. 16655883
Click to view available options
Quantity:
2.5 L
200 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
CAS Max % 1
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1013 hPa)
Quantity 200 L
Flash Point 6°C
Chemical Name or Material Acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.