missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™
196.65 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Description
Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane, 1L
- Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental and food safety industries.
- Novel gradient elution test with Photodiode Array Detector screens for UV-absorbing contaminants at every wavelength in the 200 to 400nm range to afford smooth baselines and to reduce interferences.
- Evaluated for 17 metal impurities at ppb concentrations to minimize the formation of metal adducts during analysis and simplify spectral interpretation.
- 0.1 micron filtration minimizes troublesome particles that may interfere with LC/MS and UHPLC system performance
- Provides ideal solution to prepare cleaner, particle-free mobile phase blends. Positive mode ionization (as propazine) and negative mode ionization (as mecoprop) with ESI and Single Quadrupole MS detect organic contaminants at max 50ppb and max 300ppb, respectively
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -46°C |
Absorbance | ≤1.00 AU at 190nm, ≤0.15 AU at 195nm, ≤0.05 AU at 200nm, ≤0.04 AU at 205nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.01 AU at 230nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm |
Color | Colorless |
Decomposition Information | >500°C |
Density | 0.781g/cm³ |
Boiling Point | 81°C |
Quantity | 1 L |
Flash Point | 12.8°C (55°F) |
Identification | Pass Test |
Packaging | Wide Neck Amber Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'