missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methyl cyanide, cyanomethane, acetonitrile, ethyl nitrile, ethanenitrile, cyanure de methyl, methanecarbonitrile, methylkyanid, methane, cyano, mecn, 2.5L

216.00 € - 825.00 €

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.053
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10055454
View Documents
Fisher Chemical
A955-500
500 mL
216.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10489553
View Documents
Fisher Chemical
A955-1
1 L
365.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10001334
View Documents
Fisher Chemical
A955-212
2.5 L
670.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10061044
View Documents
Fisher Chemical
A955-4
4 L
825.00 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane, 1L

  • Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental and food safety industries.
  • Novel gradient elution test with Photodiode Array Detector screens for UV-absorbing contaminants at every wavelength in the 200 to 400nm range to afford smooth baselines and to reduce interferences.
  • Evaluated for 17 metal impurities at ppb concentrations to minimize the formation of metal adducts during analysis and simplify spectral interpretation.
  • 0.1 micron filtration minimizes troublesome particles that may interfere with LC/MS and UHPLC system performance
  • Provides ideal solution to prepare cleaner, particle-free mobile phase blends. Positive mode ionization (as propazine) and negative mode ionization (as mecoprop) with ESI and Single Quadrupole MS detect organic contaminants at max 50ppb and max 300ppb, respectively
Specifications

Chemical Identifiers

75-05-8
1878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile

Specifications

-46°C
75-05-8
>500°C
81°C
Glass Bottle with fluorinated lining
Assay: 99.9% min by GC
C2H3N
CH3CN
1878
1648
≤0.01%
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%
GC
≤1.00 AU at 190nm, ≤0.15 AU at 195nm, ≤0.05 AU at 200nm, ≤0.04 AU at 205nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.01 AU at 230nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm
Colorless
0.781g/cm³
Pass Test
Liquid
≤0.8ppm
≤0.008meq/g
≤0.0006meq/g
97mbar at 20°C
0.36cp at 20°C
15, 69
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Formulated for UHPLC-UV
Acetonitrile
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health: 2
Flammability: 3
Instability: 0

missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.