missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetyl chloride, 97%

Product Code. p-7028466
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11485500

Brand: Thermo Scientific Alfa Aesar H25797.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13831-31-7 | C4H5ClO3 | 136.531 g/mol

Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

Solubility
Reacts with water violently.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13831-31-7
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00011535
InChI Key HZDNNJABYXNPPV-UHFFFAOYSA-N
Synonym acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920
PubChem CID 26297
IUPAC Name (2-chloro-2-oxoethyl) acetate
SMILES CC(=O)OCC(=O)Cl

Specifications

Density 1.27
Boiling Point 55°C (12 mmHg)
Flash Point 71°C (160°F)
Linear Formula CH3CO2CH2COCl
Refractive Index 1.4271
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water violently.
Formula Weight 136.53
Percent Purity 97%
Chemical Name or Material Acetoxyacetyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Acetoxyacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : 2-Chloro-2-oxoethylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.