Learn More
Acetyl bromide, 98%, Thermo Scientific Chemicals
97.08 € - 228.14 €
Chemical Identifiers
CAS | 506-96-7 |
---|---|
Molecular Formula | C2H3BrO |
Molecular Weight (g/mol) | 122.95 |
MDL Number | MFCD00000114 |
InChI Key | FXXACINHVKSMDR-UHFFFAOYSA-N |
Synonym | ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide |
PubChem CID | 10482 |
IUPAC Name | acetyl bromide |
SMILES | CC(=O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10734651
|
Thermo Scientific Acros
159401000 |
100 mL |
97.08 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10205970
|
Thermo Scientific Acros
159405000 |
500 mL |
228.14 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
506-96-7 | |
122.95 | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
10482 | |
CC(=O)Br |
C2H3BrO | |
MFCD00000114 | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
acetyl bromide |
Specifications
506-96-7 | |
100.0 | |
Colorless to Yellow | |
75°C to 77°C | |
Authentic | |
Glass bottle | |
CH3COBr | |
MFCD00000114 | |
06,9; 13,208 | |
1.66 | |
Solubility in water: reacts. Other solubilities: reacts with alcohol and acetone,soluble in ether,chloroform and benzene | |
CC(=O)Br | |
122.95 | |
122.95 | |
Fuming Liquid |
97.5 | |
-96°C | |
1.6600g/mL | |
75°C | |
97.5% min. (Argentometry) | |
C2H3BrO | |
1.4485 to 1.4505 | |
02, 174 | |
15, 77 | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
acetyl bromide | |
10482 | |
98% | |
Acetyl bromide, 98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-061-7
missing translation for 'rtecsNumber' : AO5955000
missing translation for 'tsca' : TSCA
Recommended Storage : May darken during storage
- Acetyl bromide
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.