Learn More
WTW™ Acid Capacity Cell and Reagent Test Kits
For use up to 4.3 pH.
Brand: WTW™ 252087
This item is not returnable.
View return policy
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Photometers | |
120 | |
120 Tests | |
Acid Capacity cell/reagent test kit |
Certificates | |
KS4.3 0.40 - 8.00mmol/L; 20 - 400 mg/L CaCO3 | |
Photometric |