missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Acid-Resistant Biopsy Bags, Medium

Reduce risk of small specimen loss with Epredia™ Acid-Resistant Biopsy Bags.

178.00 € - 314.00 €

Specifications

Product Type Biopsy Bag
Material Nylon, Acid-resistant
Color White
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12667946
View Documents
Epredia™
6774050
Medium
186.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
12677946
View Documents
Epredia™
6774051
Small
178.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
12687946
View Documents
Epredia™
6774052
Large
314.00 €
Pack of 250
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7 in. (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6 in. (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5 in. (75 × 85mm)
Specifications

Specifications

Biopsy Bag
Nylon, Acid-resistant
White
Videos
SDS
Documents

Documents

Product Certifications