Learn More
Acriflavine hydrochloride, Thermo Scientific Chemicals
Acriflavine hydrochloride, C27H28Cl4N6, CAS Number-69235-50-3, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acrifiavine hydrochloride, unii-1s73vw819c, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride
47.84 € - 221.10 €
Chemical Identifiers
CAS | 69235-50-3 |
---|---|
Molecular Formula | C27H27Cl3N6 |
Molecular Weight (g/mol) | 541.91 |
MDL Number | MFCD00069039 |
InChI Key | MKLTXAHQKDVBLY-UHFFFAOYSA-N |
Synonym | unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride |
PubChem CID | 53393742 |
IUPAC Name | acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride |
SMILES | Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10535501
|
Thermo Scientific Acros
346840100 |
10 g | Glass bottle |
47.84 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10545501
|
Thermo Scientific Acros
346840250 |
25 g | Glass bottle |
70.85 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10651832
|
Thermo Scientific Acros
346841000 |
100 g | Glass bottle |
221.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69235-50-3 | |
541.91 | |
MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
53393742 | |
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
C27H27Cl3N6 | |
MFCD00069039 | |
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | |
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride |
Specifications
69235-50-3 | |
Brown-Red to Orange-Red | |
Authentic | |
MFCD00069039 | |
15,115 | |
Solubility in water: 250-300g/L (20°C). Other solubilities: soluble in alcohol,practically insoluble in ether and chloroform | |
Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 | |
541.91 | |
Powder |
260.0°C | |
7% max. (105°C, 1 hr.) | |
C27H27Cl3N6 | |
22,I,65 | |
unii-1s73vw819c, acrifiavine hydrochloride, 3,6-diamino-10-methylacridine proflavine trihydrochloride chloride, acriflavinehydrochloride mixtureof3,6-diamino-10-methylacridiniumchloridehydrochlorideand3,6-diaminoacridinehydrochloride | |
MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;trihydrochloride | |
53393742 | |
Acriflavine hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed s
GHS Signal Word: Danger
- Acriflavine hydrochloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.