Learn More
Cytiva (formerly Pall Lab) AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
Brand:  Cytiva (formerly Pall Lab) 12995
 View more versions of this product
Product Code. 17144251
  • 215.00 € / Each
Explore more special offers
Description
Description
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrophilic PES
Sterile
1/Ea.
0.2/0.2 μm
500 cm2
Stepped Hose Barb