Learn More
Cytiva AcroPak™ 800 Capsules with Fluorodyne™ II Membrane
AcroPak 800 Capsules with Fluorodyne II (PVDF) membrane that is compatible with aqueous and many organic solvents.
Brand:  Cytiva 12471
 View more versions of this product
Product Code. 17144241
  • 218.00 € / Each
Explore more special offers
Description
Description
  • Double layer sterilizing membrane enhances reliability
  • Upstream air vent prevents vapor lock
  • Manufactured without the use of adhesives to minimize extractables
  • Pre-sterilized by gamma irradiation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hydrophilic PVDF
Sterile
Stepped Hose Barb
0.2 μm
800 cm2
1/Ea.