missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration, Long Tip
Optimized for a variety of applications requiring larger per well volume processing.
220.00 € - 310.00 €
Product Code | Brand | Pore Size | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Volume (Metric) | Price | Quantity & Availability | ||||
17124641
|
Cytiva (formerly Pall Lab)
8586 |
0.2 μm | 350 μL |
310.00 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
||||
17134641
|
Cytiva (formerly Pall Lab)
8588 |
0.45 μm | 350 μL |
310.00 €
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
17164641
|
Cytiva (formerly Pall Lab)
8686 |
0.2 μm | 1 mL |
220.00 €
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
||||
17174641
|
Cytiva (formerly Pall Lab)
8688 |
0.45 μm | 1 mL |
220.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
wwPTFE | |
Non-sterile |