missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
1083.75 EUR valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
30 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 30 kg |
Type | Silica Gel |
Material | Silica Gel |
For Use With (Application) | Column Chromatography |
Molecular Formula | O2Si |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
SMILES | O=[Si]=O |
IUPAC Name | silanedione |
Molecular Weight (g/mol) | 60.08 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'