missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide: Bis-Acrylamide 19:1 (40% Solution/Electrophoresis), Fisher BioReagents

Product Code. 10214963
Click to view available options
Quantity:
1 L
Packaging:
Polypropylene Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
This Acrylamide Solution is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
CAS 79-06-1, 110-26-9, 7732-18-5
Conductivity 10μmhos max.
Color Colorless
Quantity 1 L
DNase DNase free
Molecular Formula C3H5NO
MDL Number MFCD00008032
InChI Key HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES NC(=O)C=C
IUPAC Name prop-2-enamide
Molecular Weight (g/mol) 71.08
PubChem CID 6579
ChEBI CHEBI:28619
Grade DNase-, RNase- and Protease-Free
Packaging Polypropylene Bottle
Chemical Name or Material Acrylamide: Bis-Acrylamide 19:1, 40% Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acrylamide : Bis-Acrylamide 19:1 (40% Solution/Electrophoresis)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'