missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide/Bisacrylamide 29:1, powder

Premixed powder for preparing protein DNA gels

Brand:  Thermo Scientific Chemicals J60824.17

 View more versions of this product

Product Code. 15443529

  • 44.92 € / 40g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

Solubility
It is soluble in water.

Notes
Keep away from light. Store at room tempeature.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Acrylamide/Bisacrylamide 29:1
White
MFCD00080848
It is soluble in water.
Powder
40 g
UN2074
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only