Learn More
Thermo Scientific Chemicals Acrylamide/Bisacrylamide Solution (40%, 37.5:1)
Acrylamide/Bisacrylamide Solution (40%, 37.5:1), also known as bioAcryl-P (40%, 37.5:1), is a premixed solution containing acrylamide and N,N'-methylenebisacrylamide (in a ratio of 37.5:1, respectively) used for preparing protein DNA gels.
131.66 € - 229.14 €
Specifications
Chemical Name or Material | Acrylamide/Bisacrylamide 37.5:1 |
---|---|
Physical Form | Liquid |
Concentration | 40% soln |
Color | Colorless |
MDL Number | MFCD00080848 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15433569
|
Thermo Scientific Chemicals
J60868.AP |
500 mL |
131.66 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443569
|
Thermo Scientific Chemicals
J60868.K2 |
1 L |
229.14 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acrylamide/Bisacrylamide premixed solution is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Acrylamide/Bisacrylamide solution is used to to separate proteins
• The ratio of bisacrylamide to acrylamide modulates the porosity of the polyacrylamide gel
Application
• Acrylamide/Bisacrylamide Solution is used as a DNA sequencing gel promoting protein separation in the electrophoretic gels
• This solution is also applied for preparing gels using stock solutions with the most common cross-link ratios
• It has also been used as a precursor for polyacrylamide lid fabrication by photopatterning methods
Specifications
Acrylamide/Bisacrylamide 37.5:1 | |
40% soln | |
MFCD00080848 | |
It is soluble in water. |
Liquid | |
Colorless | |
UN3426 |
Safety and Handling
GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H340-H350-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Acrylamide/Bisacrylamide Solution (40%, 37.5:1)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide
RUO – Research Use Only