Learn More
Thermo Scientific Chemicals Acrylamide, electrophoresis grade, 99+%
Description
Acrylamide is used as a monomer in the synthesis of polyacrylamides, which finds application as water-soluble thickeners in waste water treatment, gel electrophoresis, ore processing and dyes production. In molecular biology, it is used as a carrier as well as involved in the precipitation of small amounts of DNA. It plays an important role as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 82°C to 85°C |
Density | 1.322 |
Boiling Point | 125°C (25 mmHg) |
Flash Point | 138°C (280°F) |
Odor | Odorless |
Linear Formula | CH2=CHCONH2 |
Quantity | 500 g |
UN Number | UN2074 |
Beilstein | 605349 |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
RUO – Research Use Only