Learn More
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in
Brand: Thermo Scientific Chemicals 330201000
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistrySpecifications
(1% in water), (1% in water) at 0nm, 0.1 max. at 290nm | |
Liquid | |
79-06-1, 7732-18-5, 110-26-9, 79-06-1 | |
61.0 | |
Colorless | |
Glass Bottle | |
MFCD00008032 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 |
Acrylamide / N, N'-Methylenebisacrylamide 19:1 | |
0.001% max. | |
59.0 | |
3μS/cm Max. | |
100 mL | |
C3H5NO | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
Biochemical |
Safety and Handling
- Acrylamide / N,N``-Methylenebisacrylamide 19:1, 40% mix solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only