Learn More
Brady™ Acrylic Retro Reflective Sheeting Marking Tape
The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse. This Acrylic Retro Reflective Sheeting Marking Tape gives an extra margin of safety during blackouts, at night, or in low-light conditions.
Brand:  Brady™ 076310
 View more versions of this product
Product Code. 11771925
  • 41.40 € / 1 roll
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse.
  • These tapes give an extra margin of safety during blackouts, at night, or in low-light conditions.
Specifications
Specifications
B957 2 in. X5YDS B/W STRIPED REFLECTIVE TAPE
Reflective Stripes Tape - Black / White
Semi-gloss
15 ft.
1 Roll
-40°C to +70°C
2 in.
Permanent Acrylic
Plastic
ASTM D 2979, ASTM D 523
4.57 m x 50.80 mm
Utility plant markings, Reflective Bradystake Warning Stakes, Reflective warning stripes
4.57 m
-40°F to +158°F
0.081 mm
50.8 mm
Black and White Striped
Floor Marking Tape