Learn More
Actin Gamma 1 Antibody (NH3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB100-64792
 View more versions of this product
Product Code. 18765613
  • 498.00 € / 0.25mg
Description
Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
F-Actin
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:500, Flow Cytometry 1:10, ELISA 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20
Mouse
IgM purified
RUO
71
Human, Mouse, Rat, Rabbit
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
NH3
Unconjugated
P63261
ACTG1
Human monocytes and U937 cell line
0.25 mg
Primary
Recognizes human Filamentous actin (F-actin). The binds to the N-terminal region of actin, but not to the extreme N-terminal 40 amino acids. In tissue sections the stains the cytoplasm of macrophages strongly, and gives granular, localized nuclear staining of all cell types.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgM