missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actinomycin D, Tocris Bioscience™
Click to view available options
10mg
Description
Actinomycin D is an anti-neoplastic antibiotic. Inhibits RNA polymerase and is a potent inducer of apoptosis.
Specifications
Specifications
Type | Actinomycin D |
CAS | 50-76-0 |
Molecular Formula | C62H86N12O16 |
Synonym | 2-Amino-(N,N)-1-bis(hexadecahydro-6,13-diisopropyl-2,5,9-trimethyl-1,4,7,11,14-pentaoxo-1H-pyrrolo[2,1]-[1,4,7,10,13] oxatetraazacyclohexadecin-10-yl)-4,6-dimethyl-3-oxo-3H-phenoxazine-1,9-dicarboxamide |
InChI Key | RJURFGZVJUQBHK-HUZQGMAJSA-N |
SMILES | CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |
IUPAC Name | 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
Molecular Weight (g/mol) | 1255.438 |
PubChem CID | 131954673 |
Formula Weight | 1255.43 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'