missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves
Click to view available options
Size:
7
9
Description
- Ideal for keeping hands warm in food processing and service applications
- Seamless knit, thermal acrylic liner provides comfort and warmth in cold or refrigerated work environments
- Uncoated liner can be used alone, or as an underglove
Specifications
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant |
Color | Blue |
Size | 7 |
EN388 Standard | 113XB |
For Use With (Application) | Food Processing |
Liner Material | Acrylic |
Series | 78-102 |
Yarn Gauge | 10 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction