missing translation for 'onlineSavingsMsg'
Learn More
ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm Product Code.: 16835250
Add to basket

ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm

Product Code. 16835250 Shop All Ansell Products
9
Click to view available options
Size:
7
9
This item is not returnable. View return policy

Product Code. 16835250

Brand: ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm 78202090

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Seamless knit gloves with a thermal acrylic liner to provide comfort and warmth in cold or refrigerated work environments. Micro-dot coated palm increases abrasion resistance and enhances grip capabilities.

  • Ideal for keeping hands warm in food processing and service applications
  • Seamless knit, thermal acrylic liner provides comfort and warmth in cold or refrigerated work environments
  • Uncoated liner can be used alone, or as an underglove
  • Micro-dot coating on the palm enhances grip capabilities
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH Compliant
Coating Material PVC
Color Blue
Hand Specific
Size 9
EN388 Standard 312XB
For Use With (Application) Food Processing
Liner Material Acrylic
Series 78-202
Yarn Gauge 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm ActivArmr™ 78-102 Food-Safe Thermal Insulating Gloves with Micro-Dot Palm >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.