missing translation for 'onlineSavingsMsg'
Learn More
Ansell ActivArmr™ Gloves 80-100 Product Code.: 17871692
Add to basket

Ansell ActivArmr™ Gloves 80-100

Product Code. 17871692 Shop All Ansell Products
6
Click to view available options
Size:
6
This item is not returnable. View return policy

Product Code. 17871692

Brand: Ansell 80100060

Add to basket
Add to basket
This item is not returnable. View return policy

ActivArmr™ Gloves 80-100 are durable multi-purpose gloves, combining mechanical performance with excellent grip.

  • Strengthened grip: ActivArmr™ 80-100 work gloves possess a crinkle finish, for excellent wet and dry grip, ensuring they are a safe, productivity-enhancing choice for construction, transport, shipping and warehousing applications
  • Improved durability: Robustness and durability are assured by these industrial gloves' sturdy natural rubber latex coating
  • Increased comfort: Featuring an automatic knit liner that delivers an optimally tensioned inner surface, ActivArmr™ 80-100 multi-purpose gloves ensure a comfortable fit and heightened dexterity
  • Genuine versatility: They make good work gloves as they are designed for a wide range of handling and carrying applications
  • For Seizing rough blocs and cylinders, Seizing and finishing structural parts, Loading and unloading rough parts or boxes, Picking parts and accessories, Final assembly, Loading raw material, Unloading worked parts
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance (Europe) EN 388:2003, CE Number, EN 420:2003 + A1:2009, ANSI ABR 4, ANSI Cut A2, EN 388:2016 +A1:2018, EN ISO 21420:2020, Category II, REACH Compliant
Coated Area Palm
Construction Knit
Coating Material Natural Latex
Color Blue, Gray
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Ambidextrous
Latex Free No
Material Cotton, Polyester, Natural Latex Rubber
Product Type Glove
Size 6
For Use With (Application) Construction, Transport, Shipping, Warehousing Applications
Glove Cuff Color Gray
Glove Cuff Style Knit
Liner Material Cotton, Polyester
Puncture Rating ANSI/ISEA 105-16 Level 2
Length (English) 9.25 to 11.02 in.
Length (Metric) 235 to 280 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell ActivArmr™ Gloves 80-100 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.