missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated charcoal, iodinated, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
Specifications
Specifications
Color | Black |
Packaging | Glass bottle |
Quantity | 250 g |
Merck Index | 14, 1807 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 12 |
Percent Purity | 5%, exact strength on the certificate of analysis |
Physical Form | Thin Rods |
Chemical Name or Material | Activated charcoal |
Safety and Handling
Product Identifier
- Activated charcoal, iodinated
- Warning
- Flammable solid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction