Learn More
Activator Reagent 0.5 M ETT in Acetonitrile, BioSyn™, Honeywell Burdick & Jackson™
BIOPHARMA
BioSyn™ Honeywell Burdick & Jackson™ reagent solvents for DNA/RNA synthesis
Brand:  Honeywell Burdick & Jackson BR725-4X2.5GL
Product Code. 16845431
  • 14004.41 € / Pack of 4
Explore more special offers
Description
Description
  • Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of application
  • All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Activator reagent
−46°C
Colorless
8.58% (V)
GL45 Glass Bottle
BioSyn™
2.5
75-05-8
0.92
4 x 2.5 L
Clear liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Boiling and melting point temperatures are based off the physical main component.