missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activator Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile
Specifications
CAS | 89797-68-2, 75-05-8 |
---|---|
CAS Max % | 4.00%, 96.00% |
Boiling Point | 82°C.The physical data is that of the main component. |
Flash Point | 7°C |
Physical Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825431
|
Honeywell Burdick & Jackson
BR721-4X2.5L |
4 x 2.5 L |
N/A
|
N/A | |||||
Description
Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
89797-68-2, 75-05-8 | |
82°C.The physical data is that of the main component. | |
Liquid | |
5-(Ethylthio)-1H-Tetrazole, 5-(Ethylthio)Tetrazole Solution |
4.00%, 96.00% | |
7°C | |
Activator reagents |
Safety and Handling
Recommended Storage : Room Temperature