Learn More
Memmert™ Active Microprocessor Control
Memmert™ Active Microprocessor Control ensures more rapid reaching of set humidity and very short recovery times while avoiding condensate formation.
Brand:  Memmert™ ICO150medK7
Product Code. 15968022
  • 1570.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Humidity supply with distilled water (from an external tank) by a self-priming pump
  • Integral bacteria block by generating hotsteam
  • Dehumidifying via sterile filter
Specifications
Specifications
40 - 97 % rh
40 to 97%
Humidifying and dehumidifying
ICO150MED
Humidity Supply
Digital indication and auto-diagnostic system