Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Classic and universal, these micropipettes are intended for all applications and come in a select three-pack. Socorex™ Acura™ Manual 825 Micropipette Triopack micropipettes are designed for comfort and sturdiness.
856.00 € - 913.00 €
Specifications
Certifications/Compliance | EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices |
---|---|
Autoclavable | Autoclavable |
Product Line | Acura |
Type | Micropipette Triopack |
Quantity | 3/Pk. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10680833
|
Socorex™
825.TRIO.P |
10, 100, 1000 μL |
861.00 €
1 set |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
10112412
|
Socorex™
825.TRIO.M |
10, 50, 200 μL |
856.00 €
1 set |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||||
10227032
|
Socorex™
825.TRIO.O |
0.5 to 10, 10 to 100, 100 to 1000 μL |
877.00 €
1 set |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10690833
|
Socorex™
825.TRIO.S |
2 to 20, 20 to 200, 100 to 1000 μL |
856.00 €
1 set |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
10462812
|
Socorex™
825.TRIO.T |
5 to 50, 20 to 200, 100 to 1000 μL |
856.00 €
1 set |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
10315622
|
Socorex™
825.TRIO.V |
10 to 100, 100 to 1000 μL, 0.5 to 5.0 mL |
887.00 €
1 set |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||||
10254312
|
Socorex™
835.TRIO.Y |
100 to 1000 μL, 0.5 to 5.0, 1.0 to 10 mL |
913.00 €
1 set |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||||
10142362
|
Socorex™
825.TRIO.C |
2, 10, 50 μL |
897.00 €
1 set |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
10581003
|
Socorex™
825.TRIO.J |
2, 20, 200 μL |
877.00 €
1 set |
Estimated Shipment: 10-03-2025
Log in to see stock. |
|||||
Description
- Ergonomic shape.
- Lightweight.
- Uultra-soft plunger stroke.
- Smart and reliable volume adjustment:
- State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
- Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
- Digital display of set volume on the front of the instrument is easily visible during pipetting.
- Autoclavable.
Specifications
EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices | |
Acura | |
3/Pk. |
Autoclavable | |
Micropipette Triopack |