Learn More
Socorex™ Acura™ Manual 855 Multi-channel Micropipettes
Multi-channel micropipette for use with microplates. Socorex™ Acura™ Manual 855 Multi-channel Micropipettes combine high-tech materials with exceptional ergonomics to guarantee outstanding performance and results.
Brand:  Socorex™ 855.08.010
 View more versions of this product
Product Code. 10707924
Description
Description
  • Exceptional, user-friendly ergonomics.
  • Smart and reliable volume adjustment.
  • Precision digital display visible at all times.
  • Justip™ adjustable tip ejector fitting most tips.
  • Swift-set user calibration system.
  • Shock, UV-light and autoclaving resistance.
  • 360° rotation of volumetric module allows selection of optimal working position.
  • Adjustable tip ejector.
Specifications
Specifications
<±3.5% - <±1.5%
Autoclavable
Acura™ Manual 855
Acura™
0.5 to 10 μL
1 Each
ISO 8655, GMP, GLP, NCCLS, CE directive IVD 98/79 EEC (in vitro diagnostic medical devices).
Microplate
8
Multi-Channel Pipette
<±3.0% - <±1.0%
36 Months
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products