Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand: Socorex™ 825.0010
This item is not returnable.
View return policy
Description
- Constructed of polypropylene
- With precision setting mechanism
- Single-handed volume setting
Specifications
Autoclavable | |
835 | |
Acura™ | |
0.5 to 10 μL |
Polypropylene | |
1 | |
Micropipette | |
1 Each |