missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura Variable Volume Manual Pipettes

Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.

Brand:  Socorex™ 835.10

 View more versions of this product

Product Code. 10531452

  • 349.00 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of polypropylene
  • With precision setting mechanism
  • Single-handed volume setting
Specifications

Specifications

Autoclavable
835
Acura
1 to 10 mL
Polypropylene
1
Adjustable Volume Macropipette
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Socorex™ Acura Variable Volume Manual Pipettes > Capacity: 1 to 10mL; Color: Red

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.