missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals ADA, 99%, for biochemistry
26.40 € - 69.00 €
Chemical Identifiers
CAS | 26239-55-4 |
---|---|
Molecular Formula | C6H10N2O5 |
Molecular Weight (g/mol) | 190.155 |
MDL Number | MFCD00008031 |
InChI Key | QZTKDVCDBIDYMD-UHFFFAOYSA-N |
Synonym | N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid |
PubChem CID | 117765 |
ChEBI | CHEBI:43960 |
IUPAC Name | 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
SMILES | C(C(=O)N)N(CC(=O)O)CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10145140
|
Thermo Scientific Chemicals
228010250 |
25 g | Glass bottle |
26.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321044
|
Thermo Scientific Chemicals
228011000 |
100 g | Glass bottle |
69.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
26239-55-4 | |
190.155 | |
QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
117765 | |
2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
C6H10N2O5 | |
MFCD00008031 | |
N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid | |
CHEBI:43960 | |
C(C(=O)N)N(CC(=O)O)CC(=O)O |
Specifications
(9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm | |
26239-55-4 | |
98.5 | |
White | |
99% | |
0.5% max. (110°C, 5 hrs) | |
H2NCOCH2N(CH2CO2H)2 | |
15, 135 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform | |
C(C(=O)N)N(CC(=O)O)CC(=O)O | |
190.155 | |
CHEBI:43960 | |
99% | |
ADA, For biochemistry |
(9% soln. in 1M NaOH) | |
219.0°C | |
100.0 | |
Fine Crystalline Powder | |
Authentic | |
C6H10N2O5 | |
MFCD00008031 | |
N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid | |
QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid | |
117765 | |
190.16 | |
Biochemical |
Safety and Handling
Product Identifier
- ADA
RUO – Research Use Only