missing translation for 'onlineSavingsMsg'
Learn More

Adamantane, 99+%, Thermo Scientific Chemicals

Product Code. 10091510
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281-23-2
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00074719
InChI Key ORILYTVJVMAKLC-UHFFFAOYSA-N
Synonym tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
PubChem CID 9238
ChEBI CHEBI:40519
IUPAC Name adamantane
SMILES C1C2CC3CC1CC(C2)C3

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 205.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15,137
Solubility Information Solubility in water: insoluble. Other solubilities: 8% in light petroleum fract. (20°C),6.7% in diethyl ether-1.7% in ethyl acetate (20°C),9.1% in methylene chloride-0.8% in acetone (20°C),10% in chloroform-12.5% in ccl4 (20°C),0.5%
Formula Weight 136.24
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Adamantane, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'