missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Adapta™ Universal Kinase Assay Kit

Product Code. 10678113
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10678113

Brand: Invitrogen™ PV5099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adapta™ Universal Kinase Assay Kit

The Adapta™ Universal Kinase Assay Kit is a homogenous, fluorescence-based immunoassay for the detection of ADP produced by kinases or other ATP dependent enzymes.

For more information, please visit www.invitrogen.com/adapta.
TRUSTED_SUSTAINABILITY

Specifications

Assay Universal Kinase Assay
Assay Entry Biochemical ADP accumulation
Content And Storage Adapta™ Universal Kinase Assay Kits are shipped on dry ice, and should be stored at -20°C.
Detection Method Fluorescence
Format 384-well Plate
Type Kinase Assay Kit
Excitation/Emission 340⁄615, 665
For Use With (Application) TR-FRET
Gene ID (Entrez) 1022, 1147, 120892, 1612, 3376, 5289, 5290, 5294, 8536
Label or Dye Alexa Fluor™ 647, Europium
Product Line Adapta
Quantity 1 kit
Readout End Point
Shipping Condition Dry Ice
Target Entry CAMK1, PIK3C2B, PIK3C3, PIK3CG, SPHK2, PI4KB, PIK3C2A, GSG2, LRRK2, CDK7/cyclin H/MNAT1, PI4KA, LRRK2 G2019S, CDK9/Cyclin T1, PIK3CA/PIK3R1, SPHK1, PIK3CD/PIK3R1, DAPK1
Target Enzyme Kinases (Multiple)
Validated Kinases CAMK1, CDK7, CDK9, CHUK, LRRK2, PI4KB, PIK3C2B, PIK3CA, PIK3CB, PIK3CD, PIK3CG, PIK3C2A
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.