Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.
Brand:  Eppendorf™ 5920761005
 View more versions of this product
Product Code. 16222411
  • 320.28 € / Pack of 2
Explore more special offers
Description
Description
  • Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
  • Set of 2 pieces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 x Eppendorf Tubes™ 5.0 mL and 15 mL conical tubes. (Universal adapters required for 5 mL tubes.)
For 250 mL rectangular bucket in Rotor FA-6x250
4
Set of 2 pieces