missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.
Brand: Eppendorf™ 5920767003
This item is not returnable.
View return policy
Description
- Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
- Set of 2 pieces

Specifications
3 x 30 mL Oak Ridge Round Centrifuge Tubes | |
For 250 mL rectangular bucket in Rotor FA-6x250 | |
3 | |
Set of 2 pieces |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction