missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter
Adaptors available for a variety of Eppendorf products to suit your needs.
Brand: Eppendorf™ 5920703005
315.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Adapter 500mL wide-neck bottles/RB (2x)Specifications
for 500mL Nalgene | |
Set of 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction